ENA292596460 Enamine

2-isothiocyanatobutane

  • MDL number MFCD00041108
  • Cas Number : 4426-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62130

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62130
Availability details for
Quantity
  • Availability
  • Add to Cart