ENA292596467 Enamine

tert-butyl N-(1-cyano-1-ethylpropyl)carbamate

  • MDL number MFCD12792982
  • Cas Number : 1235441-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61472
Availability details for
Quantity
  • Availability
  • Add to Cart