ENA292596468 Enamine

3-methyl-[1,2]thiazolo[5,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD10580838
  • Cas Number : 949685-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179...181
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62164

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62164
Availability details for
Quantity
  • Availability
  • Add to Cart