ENA292596470 Enamine

3-[(4-methylphenyl)sulfamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD09455231
  • Cas Number : 944894-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62167
Availability details for
Quantity
  • Availability
  • Add to Cart