ENA292596475 Enamine

2-(2-methylpiperidin-1-yl)pyridine-4-carboxylic acid

  • MDL number MFCD11131978
  • Cas Number : 1019127-10-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62174

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62174
Availability details for
Quantity
  • Availability
  • Add to Cart