ENA292596478 Enamine

(5-phenyl-1,2,4-oxadiazol-3-yl)methanol

  • MDL number MFCD04111153
  • Cas Number : 73217-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 80...82
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62179

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62179
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart