ENA292596480 Enamine

2-(2-cyclohexylacetamido)propanoic acid

  • MDL number MFCD11170032
  • Cas Number : 1397009-48-1
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62181

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62181
Availability details for
Quantity
  • Availability
  • Add to Cart