ENA292596483 Enamine

3-chloro-4-(pyridin-4-yloxy)aniline

  • MDL number MFCD11650979
  • Cas Number : 630125-41-6
  • Molecular Weight : 220.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62186
Availability details for
Quantity
  • Availability
  • Add to Cart