ENA292596484 Enamine

N-[3-(hydroxymethyl)phenyl]benzamide

  • MDL number MFCD13238529
  • Cas Number : 80936-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62187
Availability details for
Quantity
  • Availability
  • Add to Cart