ENA292596485 Enamine

2-(pyridin-2-ylsulfanyl)pyridin-4-amine

  • MDL number MFCD16040137
  • Cas Number : 1235441-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62189

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62189
Availability details for
Quantity
  • Availability
  • Add to Cart