ENA292596492 Enamine

1-[(2-fluorophenyl)methyl]-1,4-diazepane

  • MDL number MFCD03407493
  • Cas Number : 775561-26-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 50-52
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62196

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62196
Availability details for
Quantity
  • Availability
  • Add to Cart