ENA292596493 Enamine

3-(4-methoxyphenyl)pyrrolidine

  • MDL number MFCD09027609
  • Cas Number : 91246-26-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62198

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-62198
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart