ENA292596495 Enamine

1-cyclopropylethane-1,2-diamine

  • MDL number MFCD12109300
  • Cas Number : 1156284-81-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62200

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62200
Availability details for
Quantity
  • Availability
  • Add to Cart