ENA292596496 Enamine

(cyclopropylmethyl)(1-phenylethyl)amine

  • MDL number MFCD03211314
  • Cas Number : 356539-54-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62201
Availability details for
Quantity
  • Availability
  • Add to Cart