ENA292596509 Enamine

2-(3-methylbutanoyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

  • MDL number MFCD09051710
  • Cas Number : 1103531-38-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62231
Availability details for
Quantity
  • Availability
  • Add to Cart