ENA292596511 Enamine

4-(3-chlorophenoxy)benzoic acid

  • MDL number MFCD11122181
  • Cas Number : 1145-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132...134
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62233

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62233
Availability details for
Quantity
  • Availability
  • Add to Cart