ENA292596517 Enamine

2-(5-chloro-2,3-dioxo-2,3-dihydro-1H-indol-1-yl)-N-(prop-2-yn-1-yl)acetamide

  • MDL number MFCD12448422
  • Cas Number : 1020956-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62241

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62241
Availability details for
Quantity
  • Availability
  • Add to Cart