ENA292596518 Enamine

4-bromo-N-cyclopropyl-1H-pyrrole-2-carboxamide

  • MDL number MFCD12145178
  • Cas Number : 1155126-87-6
  • Molecular Weight : 229.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62242

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62242
Availability details for
Quantity
  • Availability
  • Add to Cart