ENA292596520 Enamine

3-(3,5-difluorophenyl)-1,2-oxazol-5-amine

  • MDL number MFCD11156290
  • Cas Number : 1020996-82-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62245

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62245
Availability details for
Quantity
  • Availability
  • Add to Cart