ENA292596521 Enamine

3-(2-methanesulfonylethoxy)aniline

  • MDL number MFCD12806082
  • Cas Number : 1184567-84-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62247
Availability details for
Quantity
  • Availability
  • Add to Cart