ENA292596528 Enamine

2-methyl-3-(thiomorpholine-4-carbonyl)aniline

  • MDL number MFCD11172031
  • Cas Number : 1042798-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61477
Availability details for
Quantity
  • Availability
  • Add to Cart