ENA292596532 Enamine

6-amino-1-[(4-methylphenyl)methyl]-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD06260113
  • Cas Number : 638137-05-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62269
Availability details for
Quantity
  • Availability
  • Add to Cart