ENA292596534 Enamine

2-(4-aminophenyl)-1-(thiomorpholin-4-yl)ethan-1-one

  • MDL number MFCD11172029
  • Cas Number : 1042798-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61480
Availability details for
Quantity
  • Availability
  • Add to Cart