ENA292596538 Enamine

4-(propan-2-yloxy)benzene-1-carbothioamide

  • MDL number MFCD09950634
  • Cas Number : 144060-89-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61483

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61483
Availability details for
Quantity
  • Availability
  • Add to Cart