ENA292596545 Enamine

4-{[2-(pyrrolidin-1-yl)pyridin-3-yl]carbamoyl}butanoic acid

  • MDL number MFCD16040149
  • Cas Number : 1183368-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62288

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62288
Availability details for
Quantity
  • Availability
  • Add to Cart