ENA292596547 Enamine

1-[(4-aminophenyl)methyl]piperidine-3-carboxamide

  • MDL number MFCD09729538
  • Cas Number : 953735-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62290

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62290
Availability details for
Quantity
  • Availability
  • Add to Cart