ENA292596550 Enamine

N-(3-aminopropyl)-3-hydroxybenzamide

  • MDL number MFCD09933075
  • Cas Number : 1016770-44-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62293
Availability details for
Quantity
  • Availability
  • Add to Cart