ENA292596551 Enamine

methyl 4-(3-methylpiperazin-1-yl)benzoate

  • MDL number MFCD11110714
  • Cas Number : 1131622-65-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-59406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59406
Availability details for
Quantity
  • Availability
  • Add to Cart