ENA292596553 Enamine

1-(4-methoxyphenyl)-2-(4-methylpiperazin-1-yl)ethan-1-amine

  • MDL number MFCD09731380
  • Cas Number : 953905-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62297
Availability details for
Quantity
  • Availability
  • Add to Cart