ENA292596554 Enamine

1,2,3,4-tetrahydroisoquinoline-2-carboxamide

  • MDL number MFCD09737821
  • Cas Number : 70746-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170...172
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62298

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62298
Availability details for
Quantity
  • Availability
  • Add to Cart