ENA292596555 Enamine

3-methylpiperidine-1-carboxamide

  • MDL number MFCD09732885
  • Cas Number : 953756-42-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62299

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62299
Availability details for
Quantity
  • Availability
  • Add to Cart