ENA292596556 Enamine

2-fluoro-6-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]aniline

  • MDL number MFCD12149695
  • Cas Number : 1179713-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62301

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62301
Availability details for
Quantity
  • Availability
  • Add to Cart