ENA292596562 Enamine

1-(4-aminophenyl)-N-methyl-1H-pyrazole-3-carboxamide

  • MDL number MFCD12177640
  • Cas Number : 1155164-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62310

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62310
Availability details for
Quantity
  • Availability
  • Add to Cart