ENA292596563 Enamine

2-(5-bromo-2-fluorophenyl)-2-(methylamino)acetonitrile

  • MDL number MFCD10693339
  • Cas Number : 1152577-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62311

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62311
Availability details for
Quantity
  • Availability
  • Add to Cart