ENA292596564 Enamine

4-amino-3-(prop-2-en-1-yl)-2-sulfanylidene-2,3-dihydro-1,3-thiazole-5-carbothioamide

  • MDL number MFCD03508107
  • Cas Number : 130089-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 179-182
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-59471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-59471
Availability details for
Quantity
  • Availability
  • Add to Cart