ENA292596565 Enamine

(1R)-1-(4-tert-butylphenyl)ethan-1-ol

  • MDL number MFCD09863690
  • Cas Number : 101325-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62315

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62315
Availability details for
Quantity
  • Availability
  • Add to Cart