ENA292596567 Enamine

2-(difluoromethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD08706422
  • Cas Number : 77798-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41...43
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62319

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62319
Availability details for
Quantity
  • Availability
  • Add to Cart