ENA292596573 Enamine

(oxan-2-yl)methanesulfonyl chloride

  • MDL number MFCD12783406
  • Cas Number : 187164-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-62326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62326
Availability details for
Quantity
  • Availability
  • Add to Cart