ENA292596575 Enamine

2-[2-(3,4-difluorophenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD12110827
  • Cas Number : 1156584-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 202...204
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61490

Safety Information

msds https://www.enaminestore.com/catalog/EN300-61490
Availability details for
Quantity
  • Availability
  • Add to Cart