ENA292596580 Enamine

3-[5-(2-iodophenyl)-1,3,4-oxadiazol-2-yl]propanoic acid

  • MDL number MFCD12829766
  • Cas Number : 1179674-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-61491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61491
Availability details for
Quantity
  • Availability
  • Add to Cart