ENA292596586 Enamine

5-bromo-2-[(4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]aniline

  • MDL number MFCD11132382
  • Cas Number : 1019441-45-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62373
Availability details for
Quantity
  • Availability
  • Add to Cart