ENA292596587 Enamine

1-cyclopentyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylic acid

  • MDL number MFCD06616316
  • Cas Number : 846562-87-6
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62374
Availability details for
Quantity
  • Availability
  • Add to Cart