ENA292596597 Enamine

2,3,4,5,6,7-hexahydro-1,3-benzothiazol-2-one

  • MDL number MFCD11632173
  • Cas Number : 57001-13-5
  • Molecular Weight : 155.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-62388

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62388
Availability details for
Quantity
  • Availability
  • Add to Cart