ENA292596598 Enamine

4-[2-chloro-4-(chlorosulfonyl)phenoxy]butanoic acid

  • MDL number MFCD09950718
  • Cas Number : 1016835-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62389

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62389
Availability details for
Quantity
  • Availability
  • Add to Cart