ENA292596601 Enamine

6-ethyl-2,3,4,5,6,7-hexahydro-1,3-benzothiazol-2-one

  • MDL number MFCD11632145
  • Cas Number : 1095231-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62392
Availability details for
Quantity
  • Availability
  • Add to Cart