ENA292596602 Enamine

6-(2-hydroxyethoxy)-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD12186930
  • Cas Number : 91116-12-0
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62393

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62393
Availability details for
Quantity
  • Availability
  • Add to Cart