ENA292596603 Enamine

3-[(5-aminopyridin-2-yl)oxy]benzamide

  • MDL number MFCD09933308
  • Cas Number : 1016778-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195...197
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62394

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62394
Availability details for
Quantity
  • Availability
  • Add to Cart