ENA292596607 Enamine

3-cyclohexylbutanoic acid

  • MDL number MFCD12065552
  • Cas Number : 4361-38-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62401

Safety Information

msds https://www.enaminestore.com/catalog/EN300-62401
Availability details for
Quantity
  • Availability
  • Add to Cart