ENA292596612 Enamine

2-{[(1-ethylpyrrolidin-2-yl)methyl]amino}-N,N-dimethylacetamide

  • MDL number MFCD11153900
  • Cas Number : 1021241-92-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-62406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62406
Availability details for
Quantity
  • Availability
  • Add to Cart