ENA292596622 Enamine

2-(3,5-difluorophenyl)ethan-1-amine

  • MDL number MFCD01529880
  • Cas Number : 311346-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-61502

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-61502
Availability details for
Quantity
  • Availability
  • Add to Cart